siRNA Details
Gene Symbol: Kank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270413.1 | NP_001257342.1 | 8 | 2035 | ||
XM_006242589.2 | XP_006242651.1 | 9 | 2440 | |||
XM_006242590.2 | XP_006242652.1 | 8 | 2295 |
