siRNA Details
Gene Symbol: Npsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106808.1 | NP_001100278.1 | 6 | 658 | ||
XM_008766049.1 | XP_008764271.1 | 5 | 901 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Npsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106808.1 | NP_001100278.1 | 6 | 658 | ||
XM_008766049.1 | XP_008764271.1 | 5 | 901 |