siRNA Details
Gene Symbol: Adamts15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106810.1 | NP_001100280.1 | 6 | 2075 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adamts15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106810.1 | NP_001100280.1 | 6 | 2075 |