siRNA Details
Gene Symbol: Ift46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024760.1 | NP_001019931.1 | 7 | 777 | ||
XM_006242930.2 | XP_006242992.1 | 8 | 916 | |||
XM_006242931.2 | XP_006242993.1 | 7 | 1057 | |||
GenBank mRNA | BC079388.1 | - | 7 | 683 | 7111667 | |
BC097954.1 | - | 7 | 777 | 7456463 |
