siRNA Details
Gene Symbol: Hykk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106823.1 | NP_001100293.1 | 5 | 1680 | ||
XM_006243072.2 | XP_006243134.1 | 5 | 1475 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hykk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106823.1 | NP_001100293.1 | 5 | 1680 | ||
XM_006243072.2 | XP_006243134.1 | 5 | 1475 |