siRNA Details
Gene Symbol: Hykk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106823.1 | NP_001100293.1 | 4 | 1192 | ||
XM_006243072.2 | XP_006243134.1 | 4 | 987 |

More Information
Other Species Targeted

Gene Ontology Categories:
