siRNA Details
Gene Symbol: Ubl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004247.1 | NP_001004247.1 | 2, 3 | 283 | ||
XM_006243155.2 | XP_006243217.1 | 2, 3 | 251 | |||
XM_006243156.2 | XP_006243218.1 | 2, 3 | 303 | |||
GenBank mRNA | BC079221.1 | - | 2, 3 | 283 | 7131980 | |
FQ209768.1 | - | 1, 2 | 240 |
