siRNA Details
Gene Symbol: Parp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106828.1 | NP_001100298.1 | 19 | 1917 | ||
XM_006243191.1 | XP_006243253.1 | 19 | 2201 | |||
XM_006243192.2 | XP_006243254.1 | 18 | 2859 | |||
XM_006243193.2 | XP_006243255.1 | 18 | 1728 | |||
XM_008766231.1 | XP_008764453.1 | 18 | 2856 | |||
XM_008766232.1 | XP_008764454.1 | 18 | 2799 | |||
XM_008766233.1 | XP_008764455.1 | 18 | 2796 |
