siRNA Details
Gene Symbol: Gramd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243197.2 | XP_006243259.1 | 5 | 602 | ||
XM_006243198.2 | XP_006243260.1 | 6 | 963 | |||
XM_006243199.2 | XP_006243261.1 | 5 | 1011 | |||
XM_006243200.2 | XP_006243262.1 | 5 | 1351 | |||
XM_006243201.2 | XP_006243263.1 | 5 | 2812 | |||
XM_008766374.1 | XP_008764596.1 | 5 | 482 | |||
XM_217153.7 | XP_217153.5 | 5 | 401 |
