siRNA Details
Gene Symbol: Iqch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106830.1 | NP_001100300.1 | 4 | 428 | ||
XM_008766237.1 | XP_008764459.1 | 4 | 484 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Iqch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106830.1 | NP_001100300.1 | 4 | 428 | ||
XM_008766237.1 | XP_008764459.1 | 4 | 484 |