siRNA Details
Gene Symbol: Iqch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106830.1 | NP_001100300.1 | 15 | 2134 | ||
XM_008766234.1 | XP_008764456.1 | 9 | 1650 | |||
XM_008766236.1 | XP_008764458.1 | 10 | 1734 | |||
XM_008766237.1 | XP_008764459.1 | 15 | 2235 |

More Information
Other Species Targeted

