siRNA Details
Gene Symbol: Bnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106835.1 | NP_001100305.1 | 4, 5 | 549 | ||
XM_006243370.2 | XP_006243432.1 | 4, 5 | 593 | |||
XM_006243371.2 | XP_006243433.1 | 4, 5 | 591 |
