siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106836.1 | NP_001100306.1 | 6 | 1649 | ||
XM_006243372.2 | XP_006243434.1 | 6 | 1802 | |||
XM_006243373.2 | XP_006243435.1 | 6 | 1801 | |||
XM_006243374.2 | XP_006243436.1 | 6 | 1801 | |||
XM_006243376.2 | XP_006243438.1 | 7 | 1894 | |||
XM_006243377.2 | XP_006243439.1 | 6 | 1831 | |||
XM_008766243.1 | XP_008764465.1 | 6 | 1801 | |||
XM_008766244.1 | XP_008764466.1 | 6 | 1580 | |||
XM_008766245.1 | XP_008764467.1 | 6 | 1802 |
