siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106836.1 | NP_001100306.1 | 2 | 763 | ||
XM_006243372.2 | XP_006243434.1 | 2 | 916 | |||
XM_006243373.2 | XP_006243435.1 | 2 | 915 | |||
XM_006243374.2 | XP_006243436.1 | 2 | 915 | |||
XM_006243376.2 | XP_006243438.1 | 3 | 1008 | |||
XM_006243377.2 | XP_006243439.1 | 2 | 945 | |||
XM_008766243.1 | XP_008764465.1 | 2 | 915 | |||
XM_008766244.1 | XP_008764466.1 | 2 | 694 | |||
XM_008766245.1 | XP_008764467.1 | 2 | 916 |
