siRNA Details
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013252.1 | NP_001013270.1 | 4 | 400 | ||
GenBank mRNA | AY325214.1 | - | 2548 | |||
AY325230.1 | - | 2551 | ||||
AY325241.1 | - | 2548 | ||||
BC098951.1 | - | 392 | 7324816 | |||
BF523222.1 | - | 1 | 66 | |||
BF525252.1 | - | 406 | ||||
BQ204036.1 | - | 4 | 511 |
