siRNA Details
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013252.1 | NP_001013270.1 | 5 | 511 | ||
GenBank mRNA | AY325214.1 | - | 2659 | |||
AY325230.1 | - | 2662 | ||||
AY325241.1 | - | 2659 | ||||
BC098951.1 | - | 503 | 7324816 | |||
BF523222.1 | - | 2 | 177 |
