siRNA Details
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013252.1 | NP_001013270.1 | 2, 3 | 258 | ||
GenBank mRNA | AY325214.1 | - | 2406 | |||
AY325230.1 | - | 2409 | ||||
AY325241.1 | - | 2406 | ||||
BC098951.1 | - | 250 | 7324816 | |||
BF525252.1 | - | 264 | ||||
BQ204036.1 | - | 5, 6 | 653 |
