siRNA Details
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008328.1 | NP_001008329.1 | 9 | 1362 | ||
XM_006243725.2 | XP_006243787.1 | 9 | 1698 | |||
XM_008766500.1 | XP_008764722.1 | 9 | 1729 | |||
GenBank mRNA | BC085863.1 | - | 9 | 1362 | 7190424 |
