siRNA Details
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100548.1 | NP_001094018.1 | 7 | 800 | ||
XM_006243738.2 | XP_006243800.1 | 7 | 1054 | |||
XM_006243739.2 | XP_006243801.1 | 7 | 692 | |||
GenBank mRNA | BC166477.1 | - | 7 | 800 | 9102173 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_148930, AK146880, AJ309168, AK134183, BC031899, and others.
Gene Ontology Categories:
