siRNA Details
Gene Symbol: Slc26a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143817.1 | NP_001137289.1 | 7 | 861 | ||
XM_006243755.2 | XP_006243817.1 | 7 | 2889 | |||
XM_006243756.2 | XP_006243818.1 | 7 | 2067 | |||
XM_006243757.2 | XP_006243819.1 | 7 | 1433 | |||
XM_006243758.2 | XP_006243820.1 | 7 | 1434 | |||
XM_008766510.1 | XP_008764732.1 | 6 | 1329 |
