siRNA Details
Gene Symbol: Slc26a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143817.1 | NP_001137289.1 | 11 | 1295 | ||
XM_006243755.2 | XP_006243817.1 | 11 | 3323 | |||
XM_006243756.2 | XP_006243818.1 | 11 | 2501 | |||
XM_006243757.2 | XP_006243819.1 | 11 | 1867 | |||
XM_006243758.2 | XP_006243820.1 | 11 | 1868 | |||
XM_008766510.1 | XP_008764732.1 | 10 | 1763 |
