siRNA Details
Gene Symbol: LOC100911807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750589.3 | XP_003750637.1 | 5, 6 | 1928 |
Gene Symbol: Atrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106859.1 | NP_001100329.1 | 5, 6 | 933 | ||
XM_006243764.2 | XP_006243826.1 | 5, 6 | 935 | |||
XM_006243766.2 | XP_006243828.1 | 5, 6 | 934 | |||
XM_006243767.2 | XP_006243829.1 | 5, 6 | 934 |
