siRNA Details
Gene Symbol: Lrrfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024761.1 | NP_001019932.1 | 10, 11 | 1058 | ||
XM_006243939.2 | XP_006244001.1 | 22, 23 | 1843 | |||
XM_006243957.1 | XP_006244019.1 | 9, 10 | 1097 | |||
XM_006243958.2 | XP_006244020.1 | 9, 10 | 1037 | |||
XM_006243960.1 | XP_006244022.1 | 8, 9 | 1025 | |||
GenBank mRNA | BC097955.1 | - | 10, 11 | 1058 | 7456701 | |
FQ233539.1 | - | 9, 10 | 1081 |
