siRNA Details
Gene Symbol: Vill | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191617.1 | NP_001178546.1 | 17 | 2192 | ||
XM_006244085.1 | XP_006244147.1 | 18 | 2291 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vill | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191617.1 | NP_001178546.1 | 17 | 2192 | ||
XM_006244085.1 | XP_006244147.1 | 18 | 2291 |