siRNA Details
Gene Symbol: Guca1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106887.1 | NP_001100357.1 | 2 | 343 | ||
XM_006244428.2 | XP_006244490.1 | 1 | 298 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Guca1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106887.1 | NP_001100357.1 | 2 | 343 | ||
XM_006244428.2 | XP_006244490.1 | 1 | 298 |