siRNA Details
Gene Symbol: Enpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106892.1 | NP_001100362.1 | 2 | 812 | ||
XM_006244598.2 | XP_006244660.1 | 3 | 2208 | |||
XM_006244600.2 | XP_006244662.1 | 2 | 2093 | |||
XM_006244601.2 | XP_006244663.1 | 4 | 1857 | |||
XM_006244602.2 | XP_006244664.1 | 2 | 794 | |||
XM_008766852.1 | XP_008765074.1 | 3 | 908 | |||
XM_008766853.1 | XP_008765075.1 | 2 | 702 |
