siRNA Details
Gene Symbol: Pla2g7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009353.1 | NP_001009353.1 | 11 | 1321 | ||
XM_006244606.2 | XP_006244668.1 | 11 | 1438 | |||
XM_008766856.1 | XP_008765078.1 | 10 | 3081 | |||
GenBank mRNA | BC088457.1 | - | 11 | 1321 | 7127300 |
