siRNA Details
Gene Symbol: Coq10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009671.1 | NP_001009671.1 | 4 | 541 | ||
XM_008767080.1 | XP_008765302.1 | 4 | 842 | |||
GenBank mRNA | BC088273.1 | - | 4 | 541 | 7380093 | |
FQ219530.1 | - | 3 | 400 |

More Information
Other Species Targeted

