siRNA Details
Gene Symbol: Tyw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170473.1 | NP_001163944.1 | 8 | 1096 | ||
XM_006244910.2 | XP_006244972.1 | 7 | 702 | |||
GenBank mRNA | BC158619.1 | - | 8 | 1077 | ||
CK845177.1 | - | 1 | 393 |
