siRNA Details
Gene Symbol: Kctd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106914.1 | NP_001100384.1 | 4 | 3685 | ||
XM_006244965.2 | XP_006245027.1 | 2 | 1734 | |||
XM_006244966.2 | XP_006245028.1 | 2 | 286 | |||
XM_006244967.2 | XP_006245029.1 | 2 | 798 | |||
XM_008767084.1 | XP_008765306.1 | 1 | 2480 | |||
XM_008767085.1 | XP_008765307.1 | 1 | 2598 | |||
XM_008767086.1 | XP_008765308.1 | 2 | 291 | |||
XM_008767087.1 | XP_008765309.1 | 2 | 220 | |||
GenBank mRNA | FQ209862.1 | - | 3 | 853 | ||
FQ228496.1 | - | 3 | 854 |
