siRNA Details
Gene Symbol: Kctd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106914.1 | NP_001100384.1 | 8, 9 | 4333 | ||
XM_006244965.2 | XP_006245027.1 | 6, 7 | 2382 | |||
XM_006244966.2 | XP_006245028.1 | 6, 7 | 934 | |||
XM_006244967.2 | XP_006245029.1 | 6, 7 | 1446 | |||
XM_008767084.1 | XP_008765306.1 | 5, 6 | 3128 | |||
XM_008767085.1 | XP_008765307.1 | 5, 6 | 3246 | |||
XM_008767086.1 | XP_008765308.1 | 6, 7 | 939 | |||
XM_008767087.1 | XP_008765309.1 | 6, 7 | 868 | |||
GenBank mRNA | FQ209862.1 | - | 7, 8 | 1479 | ||
FQ228496.1 | - | 7, 8 | 1502 |
