siRNA Details
Gene Symbol: Rnf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012004.2 | NP_001012004.2 | 3 | 183 | ||
XM_006245172.2 | XP_006245234.1 | 3 | 214 | |||
XM_006245173.1 | XP_006245235.1 | 3 | 1064 | |||
GenBank mRNA | CF114855.1 | - | 3 | 193 |
