siRNA Details
Gene Symbol: Stk36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173986.1 | NP_001167457.1 | 12 | 1776 | ||
XM_006245174.2 | XP_006245236.1 | 12 | 1789 | |||
XM_008767221.1 | XP_008765443.1 | 8 | 1198 | |||
GenBank mRNA | FQ218305.1 | - | 12 | 1778 |
