siRNA Details
Gene Symbol: Sgpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191811.1 | NP_001178740.1 | 2 | 164 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sgpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191811.1 | NP_001178740.1 | 2 | 164 |