siRNA Details
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106924.1 | NP_001100394.1 | 4 | 841 | ||
XM_006245373.2 | XP_006245435.1 | 5 | 990 | |||
XM_006245374.2 | XP_006245436.1 | 5 | 1133 | |||
XM_006245375.2 | XP_006245437.1 | 5 | 892 | |||
GenBank mRNA | FQ225617.1 | - | 1 | 157 | ||
FQ232683.1 | - | 1 | 157 |

More Information
Other Species Targeted

Gene Ontology Categories:
