siRNA Details
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106924.1 | NP_001100394.1 | 3 | 632 | ||
XM_006245373.2 | XP_006245435.1 | 3 | 662 | |||
XM_006245374.2 | XP_006245436.1 | 3 | 805 | |||
XM_006245375.2 | XP_006245437.1 | 3 | 564 |
