siRNA Details
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106924.1 | NP_001100394.1 | 5 | 888 | ||
XM_006245373.2 | XP_006245435.1 | 6 | 1037 | |||
XM_006245374.2 | XP_006245436.1 | 6 | 1180 | |||
XM_006245375.2 | XP_006245437.1 | 6 | 939 | |||
GenBank mRNA | FQ225617.1 | - | 2 | 204 | ||
FQ226825.1 | - | 1 | 30 | |||
FQ232683.1 | - | 2 | 204 | |||
FQ233663.1 | - | 1 | 29 |
