siRNA Details
Gene Symbol: RGD1304704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047874.1 | NP_001041339.1 | 3 | 268 | ||
XM_006251636.2 | XP_006251698.1 | 2 | 274 | |||
XM_006251638.2 | XP_006251700.1 | 2 | 275 | |||
GenBank mRNA | AY539943.1 | - | 3 | 268 | ||
FQ215371.1 | - | 2 | 217 | |||
FQ220810.1 | - | 2 | 261 | |||
FQ224955.1 | - | 2 | 268 | |||
FQ229372.1 | - | 2 | 271 | |||
FQ229497.1 | - | 2 | 255 |
