siRNA Details
Gene Symbol: Pmm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106973.1 | NP_001100443.1 | 7 | 612 | ||
XM_006245755.1 | XP_006245817.1 | 8 | 951 | |||
GenBank mRNA | BC168922.1 | - | 7 | 568 | 7122126 | |
FQ232490.1 | - | 2 | 106 |
