siRNA Details
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009676.1 | NP_001009676.1 | 6, 7 | 744 | ||
XM_006245788.1 | XP_006245850.1 | 6, 7 | 787 | |||
XM_006245789.2 | XP_006245851.1 | 6, 7 | 787 | |||
XM_006245790.1 | XP_006245852.1 | 6, 7 | 680 | |||
XM_006245791.1 | XP_006245853.1 | 3, 4 | 391 | |||
XM_008767482.1 | XP_008765704.1 | 6, 7 | 787 | |||
XM_008767483.1 | XP_008765705.1 | 5, 6 | 2516 | |||
XM_008767484.1 | XP_008765706.1 | 4, 5 | 391 | |||
GenBank mRNA | BC087062.1 | - | 6, 7 | 744 | 7131099 |
