siRNA Details
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009676.1 | NP_001009676.1 | 3, 4 | 426 | ||
XM_006245788.1 | XP_006245850.1 | 3, 4 | 469 | |||
XM_006245789.2 | XP_006245851.1 | 3, 4 | 469 | |||
XM_006245790.1 | XP_006245852.1 | 3, 4 | 362 | |||
XM_008767482.1 | XP_008765704.1 | 3, 4 | 469 | |||
XM_008767483.1 | XP_008765705.1 | 2, 3 | 2198 | |||
XM_008767484.1 | XP_008765706.1 | 1, 2 | 73 | |||
GenBank mRNA | BC087062.1 | - | 3, 4 | 426 | 7131099 |

More Information
Other Species Targeted

