siRNA Details
Gene Symbol: Tpsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175593.2 | NP_783183.1 | 1 | 24 | ||
GenBank mRNA | AY196208.1 | - | 1 | 24 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tpsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175593.2 | NP_783183.1 | 1 | 24 | ||
GenBank mRNA | AY196208.1 | - | 1 | 24 |