siRNA Details
Gene Symbol: Tekt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013965.1 | NP_001013987.1 | 1 | 391 | ||
GenBank mRNA | AB194013.1 | - | 1 | 276 | ||
BC079304.1 | - | 1 | 391 | 7134598 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tekt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013965.1 | NP_001013987.1 | 1 | 391 | ||
GenBank mRNA | AB194013.1 | - | 1 | 276 | ||
BC079304.1 | - | 1 | 391 | 7134598 |