siRNA Details
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030034.1 | NP_001025205.1 | 4 | 470 | ||
XM_006245977.2 | XP_006246039.1 | 4 | 4288 | |||
XM_008767578.1 | XP_008765800.1 | 5 | 3405 | |||
XM_008767579.1 | XP_008765801.1 | 5 | 3280 | |||
XM_008767580.1 | XP_008765802.1 | 4 | 1390 | |||
GenBank mRNA | BC099777.1 | - | 4 | 470 | 7933232 |
