siRNA Details
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030034.1 | NP_001025205.1 | 6, 7 | 918 | ||
XM_006245977.2 | XP_006246039.1 | 6, 7 | 4736 | |||
XM_008767578.1 | XP_008765800.1 | 7, 8 | 3853 | |||
XM_008767579.1 | XP_008765801.1 | 7, 8 | 3728 | |||
XM_008767580.1 | XP_008765802.1 | 6, 7 | 1838 | |||
GenBank mRNA | BC099777.1 | - | 6, 7 | 918 | 7933232 |
