siRNA Details
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030034.1 | NP_001025205.1 | 4, 5 | 587 | ||
XM_006245977.2 | XP_006246039.1 | 4, 5 | 4405 | |||
XM_008767578.1 | XP_008765800.1 | 5, 6 | 3522 | |||
XM_008767579.1 | XP_008765801.1 | 5, 6 | 3397 | |||
XM_008767580.1 | XP_008765802.1 | 4, 5 | 1507 | |||
GenBank mRNA | BC099777.1 | - | 4, 5 | 587 | 7933232 |
