siRNA Details
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106992.1 | NP_001100462.1 | 8, 9 | 1947 | ||
XM_006245978.2 | XP_006246040.1 | 9, 10 | 3345 | |||
XM_006245979.2 | XP_006246041.1 | 8, 9 | 3294 | |||
XM_006245980.2 | XP_006246042.1 | 7, 8 | 3270 |

More Information
Other Species Targeted

Gene Ontology Categories:
