siRNA Details
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106992.1 | NP_001100462.1 | 1 | 1085 | ||
XM_006245978.2 | XP_006246040.1 | 1 | 2459 | |||
XM_006245979.2 | XP_006246041.1 | 1 | 2459 | |||
XM_006245980.2 | XP_006246042.1 | 1 | 2459 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026252, AK173229, AY313775, BC115430, BC145865, and others.
Gene Ontology Categories:
