siRNA Details
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106993.1 | NP_001100463.1 | 4 | 452 | ||
XM_006246092.2 | XP_006246154.1 | 4 | 458 | |||
XM_006246093.2 | XP_006246155.1 | 4 | 449 | |||
XM_006246094.2 | XP_006246156.1 | 3 | 386 |
