siRNA Details
Gene Symbol: Ranbp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106994.1 | NP_001100464.1 | 11 | 1153 | ||
XM_008767582.1 | XP_008765804.1 | 11 | 2842 | |||
XM_008767583.1 | XP_008765805.1 | 11 | 2815 |
